Learn More
Medchemexpress LLC Triammonium phosphate trihydrate | 25447-33-0 | MFCD00151223 | ≥98.0% | 203.13 | H18N3O7P | 1kg
SDP
Catalog No. 5000269788
Encompass_Preferred
Triammonium phosphate trihydrate interacts with the magnesium ions to form a precipitate which can be used for magnesium separation from lithium in salt-lake brines[1]
Catalog No. 50-002-69788 Supplier Medchemexpress LLC Supplier No. HYY10051KG
Triammonium phosphate trihydrate interacts with the magnesium ions to form a precipitate which can be used for magnesium separation from lithium in salt-lake brines[1]