Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Triamterene | 396-01-0 | MFCD00006708 | 99.9% | 253.26 g/mol | C12H11N7 | 10 MG
SDP

Catalog No. 5000298075
Encompass_Preferred

Triamterene (standard) is an analytical reference material intended for use in research and analytical applications, including calibration, method development, and quality control. The material is supplied as a solid with defined molecular properties and high reported purity for reproducible assay performance.

  • Analytical reference standard for triamterene.
  • High purity of 99.9%.
  • Molecular weight 253.26 g/mol.
  • CAS number 396-01-0.
  • Available as a 10 MG solid package.

Catalog No. 50-002-98075 Supplier Medchemexpress LLC Supplier No. HYB0575R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Triamterene (standard) is an analytical reference material intended for use in research and analytical applications, including calibration, method development, and quality control. The material is supplied as a solid with defined molecular properties and high reported purity for reproducible assay performance.

  • Analytical reference standard for triamterene.
  • High purity of 99.9%.
  • Molecular weight 253.26 g/mol.
  • CAS number 396-01-0.
  • Available as a 10 MG solid package.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.