Learn More
Medchemexpress LLC Triamterene-d5 | 1189922-23-3 | 99.8% | 1 MG
SDP
Catalog No. 5000408625
Encompass_Preferred
Triamterene-d5 is a deuterium-labeled form of Triamterene, which functions by blocking the epithelial Na+ channel (ENaC) in a voltage-dependent manner, acting as a mild diuretic. It also serves as an inhibitor of the TGR5 receptor. This compound is intended for research use only and is not for sale to patients.
  • Deuterium-labeled compound
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Blocks epithelial Na+ channel (ENaC)
  • Acts as a mild diuretic
  • Inhibitor of the TGR5 receptor
Catalog No. 50-004-08625 Supplier Medchemexpress LLC Supplier No. HYB0575S1MG
Triamterene-d5 is a deuterium-labeled form of Triamterene, which functions by blocking the epithelial Na+ channel (ENaC) in a voltage-dependent manner, acting as a mild diuretic. It also serves as an inhibitor of the TGR5 receptor. This compound is intended for research use only and is not for sale to patients.
  • Deuterium-labeled compound
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Blocks epithelial Na+ channel (ENaC)
  • Acts as a mild diuretic
  • Inhibitor of the TGR5 receptor