Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triamylamine (mixture of branched chain isomers) 97.0+%, TCI America™
SDP

Catalog No. T0338500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0338500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T0338500ML Supplier TCI America Supplier No. T0338500ML

missing translation for 'validMainframeMsg'

Specifications

CAS 621-77-2
Color Yellow
Molecular Formula C15H33N
MDL Number MFCD00015270
UN Number 2735
Quantity 500 mL
Synonym triamylamine, tripentylamine, tri-n-pentylamine, 1-pentanamine, n,n-dipentyl, tri-n-amylamine, ccris 4877, tripentyl amine, 1-pentanamine,n-dipentyl, acmc-209n0e, 1-pentanamine,n,n-dipentyl
InChI Key OOHAUGDGCWURIT-UHFFFAOYSA-N
SMILES CCCCCN(CCCCC)CCCCC
IUPAC Name N,N-dipentylpentan-1-amine
Molecular Weight (g/mol) 227.436
PubChem CID 12133
Formula Weight 227.44
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Triamylamine (mixture of branched chain isomers)
Show More Show Less
EINECSNumber (2)-3526
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.