Learn More
General Data Healthcare™ TEC™-II Tissue Embedding Center: Dispensing Console
SDP
Catalog No. 22046851 Shop All Triangle Biomedical Sciences Inc. (TBS) Products
Change view
Click to view available options
Product Type:
Dispensing Console
Catalog No. Product Type
22-046-851 Dispensing Console
1 options
Catalog No. 22-046-851 Supplier General Data Healthcare™ Supplier No. TEC2120DC
For use with Triangle Biomedical Services TEC-II Tissue Embedding Center (cat. no. 22046850)
  • 5-liter paraffin reservoir
  • Heated dispenser nozzle and heated work surfaces, heated forceps holders and a cold spot
  • Includes removable base mold and cassette trays that are independently heated in their own compartments
  • 590 x 655 x 425mm (23 x 26 x 17 in.)
Specifications
Product Type Dispensing Console
Weight (English) Module 2187 oz.
Weight (Metric) Module 62 kg
Voltage 100/120 V
Frequency 50/60 Hz