Learn More
Alkali Scientific Triangle Spreader, 60 x 235mm (25/Sterile Bag, 4 Bags), 100/cs
SDP
Catalog No. 502389054
Encompass_Preferred
These triangle shape spreaders are used for CFU count applications, and have a grooved handle that improves grip and orientation. Made of polypropylene, they are available with an extra large 60mm span.
Catalog No. 50-238-9054 Supplier Alkali Scientific Supplier No. TS5441
These triangle shape spreaders are used for CFU count applications, and have a grooved handle that improves grip and orientation. Made of polypropylene, they are available with an extra large 60mm span.