Learn More
Grainger Micro100 Triangle Turning Insert, Solid Carbide, Uncoated, 3-Edge, 1 ea
Catalog No. 502778520
Encompass_Preferred
The Triangle Turning Insert TP-41 is a high-performance, solid carbide indexable cutter designed for versatile machining applications. Featuring a positive rake and a triangle shape, this insert offers reliable performance across a wide range of materials, including steel, stainless steel, cast iron, and non-ferrous metals. It is manufactured from proprietary micrograin carbide to provide excellent shock resistance and strength under demanding conditions.
  • Triangle shape with 3 cutting edges
  • Solid carbide construction for durability and shock resistance
  • Uncoated finish
  • 10° relief angle for improved cutting performance
  • Compatible with steel, stainless steel, cast iron, hardened steel, and non-ferrous materials
  • Designed for screw-on mounting with a 0.137 in. hole diameter
  • 0.0156 in. corner radius
Catalog No. 50-277-8520 Supplier Grainger Supplier No. 18D751
The Triangle Turning Insert TP-41 is a high-performance, solid carbide indexable cutter designed for versatile machining applications. Featuring a positive rake and a triangle shape, this insert offers reliable performance across a wide range of materials, including steel, stainless steel, cast iron, and non-ferrous metals. It is manufactured from proprietary micrograin carbide to provide excellent shock resistance and strength under demanding conditions.
  • Triangle shape with 3 cutting edges
  • Solid carbide construction for durability and shock resistance
  • Uncoated finish
  • 10° relief angle for improved cutting performance
  • Compatible with steel, stainless steel, cast iron, hardened steel, and non-ferrous materials
  • Designed for screw-on mounting with a 0.137 in. hole diameter
  • 0.0156 in. corner radius