Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ampco Triangular Scraper
SDP

Catalog No. 19173543 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
12 -1/2 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
19-173-543 12 -1/2 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-173-543 Supplier Ampco Supplier No. 41268
Add to Cart
Add to Cart

Unique non-sparking, non-magnetic, and corrosion-resistant triangular blade makes it versatile and safe for use in hazardous or sensitive environment.

  • Made in the United Kingdom
  • Overall length of 320 mm or 12 and 1/2 in.
  • Blade length of 200 mm or 8 in.

Specifications

Product Type Triangular Scraper
Length (English) 12 -1/2 in.
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.