Learn More
Seedburo Equipment Company Blue Powdercoat Steel Triangular Sample Pan, 10 x 10 x 2.5''
SDP
Catalog No. NC1487464
Encompass
The No. 64 Sample Pan is made of mild steel and has an enameled, dark blue color. This pan has become a standard and features a tapered pour spout.
Catalog No. NC1487464 Supplier Seedburo Equipment Company Supplier No. 64
The No. 64 Sample Pan is made of mild steel and has an enameled, dark blue color. This pan has become a standard and features a tapered pour spout.