Learn More
Medchemexpress LLC Triantennary N-acetylgalactosamine (GalNAc-L96) | 1159408-62-4 | 97.0% | C127H194N12O45 | 10MG
SDP
Catalog No. 5000258480
Encompass_Preferred
GalNAc-L96 is a triantennary N-acetylgalactosamine ligand of the asialoglycoprotein receptor (ASGPR). It is used as a targeting moiety in the synthesis of GalNAc-conjugated oligonucleotides and siRNA for nucleic acid drug delivery research.
  • Triantennary GalNAc ligand for ASGPR targeting.
  • Suitable for synthesis of GalNAc-conjugated oligonucleotides and siRNA.
  • Off-white to light yellow solid, convenient for handling.
  • Typical purity ~97.0%.
  • Molecular formula C127H194N12O45; molecular weight ~2608.95.
  • Available in small to bulk package sizes for research use.
Catalog No. 50-002-58480 Supplier Medchemexpress LLC Supplier No. HY14701210MG
GalNAc-L96 is a triantennary N-acetylgalactosamine ligand of the asialoglycoprotein receptor (ASGPR). It is used as a targeting moiety in the synthesis of GalNAc-conjugated oligonucleotides and siRNA for nucleic acid drug delivery research.
  • Triantennary GalNAc ligand for ASGPR targeting.
  • Suitable for synthesis of GalNAc-conjugated oligonucleotides and siRNA.
  • Off-white to light yellow solid, convenient for handling.
  • Typical purity ~97.0%.
  • Molecular formula C127H194N12O45; molecular weight ~2608.95.
  • Available in small to bulk package sizes for research use.