Learn More
Apexbio Technology LLC Triapine 236392-56-6; 200933-27-3; 143621-35-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000064246
Encompass_Preferred
Triapine is a small molecule thiosemicarbazone derivative known to inhibit ribonucleotide reductase an enzyme essential for the conversion of ribonucleotides to deoxyribonucleotides thereby interfering with DNA synthesis and repair pathways Through this inhibition mechanism Triapine exhibits antitumor activity in a range of experimental cancer models including human ovarian carcinoma xenografts and murine leukemia models and inhibits proliferation across multiple tumor cell lines Due to its mechanistic specificity Triapine serves as a useful chemical tool for biomedical research into nucleic acid metabolism cancer cell replication and therapeutic resistance mechanisms
Catalog No. 50-000-64246 Supplier Apexbio Technology LLC Supplier No. B32095.1
Triapine is a small molecule thiosemicarbazone derivative known to inhibit ribonucleotide reductase an enzyme essential for the conversion of ribonucleotides to deoxyribonucleotides thereby interfering with DNA synthesis and repair pathways Through this inhibition mechanism Triapine exhibits antitumor activity in a range of experimental cancer models including human ovarian carcinoma xenografts and murine leukemia models and inhibits proliferation across multiple tumor cell lines Due to its mechanistic specificity Triapine serves as a useful chemical tool for biomedical research into nucleic acid metabolism cancer cell replication and therapeutic resistance mechanisms