Learn More
Apexbio Technology LLC Triapine, 25mg. Cas: 236392-56-6; 200933-27-3; 143621-35-6 MFCD: MFCD00924747. Ribonucleotide reductase inhibitor
SDP
Supplier:  Apexbio Technology LLC B320925
Encompass_Preferred
Triapine is a potent inhibitor of ribonucleotide reductase activity with IC50 value of 1.6 M for various of tumor cell lines [1]. Triapine has been reported to inhibit ribonucleotide reductase activity. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5344