Learn More
Medchemexpress LLC Triarachidin | 620-64-4 | MFCD00042879 | 99.0% | 975.64 g/mol | C63H122O6 | 250MG
SDP
Catalog No. 5000239622
Encompass_Preferred
Triarachidin is an endogenous glycerol tri-icosanoate supplied as a research-grade lipid standard and metabolic reference. It is provided as a high-purity material for biochemical and analytical workflows, including lipidomics, method development, and instrument calibration. Documentation such as a datasheet, safety data sheet, and certificate of analysis is typically available to support experimental use.
  • High purity 99.0% suitable for analytical and research use.
  • Molecular weight 975.64 g/mol and formula C63H122O6 for accurate calculations.
  • Useful as a lipid standard and metabolic reference in lipidomics and metabolic studies.
  • Provided with datasheet, safety data sheet, and certificate of analysis.
  • Available in laboratory pack sizes for convenient handling and storage.
Catalog No. 50-002-39622 Supplier Medchemexpress LLC Supplier No. HY125671250MG
Triarachidin is an endogenous glycerol tri-icosanoate supplied as a research-grade lipid standard and metabolic reference. It is provided as a high-purity material for biochemical and analytical workflows, including lipidomics, method development, and instrument calibration. Documentation such as a datasheet, safety data sheet, and certificate of analysis is typically available to support experimental use.
  • High purity 99.0% suitable for analytical and research use.
  • Molecular weight 975.64 g/mol and formula C63H122O6 for accurate calculations.
  • Useful as a lipid standard and metabolic reference in lipidomics and metabolic studies.
  • Provided with datasheet, safety data sheet, and certificate of analysis.
  • Available in laboratory pack sizes for convenient handling and storage.