Learn More
YMC America, Inc TRIART C8 120A 3UM 150X4.6MM
SDP
Supplier:  YMC America, Inc TO12S031546WT
Encompass_Preferred
TO12S03-1546WT: Triart C8, 120A, 3um, 150x4.6mm Endcapped Column
Catalog No. 50-394-58