Learn More
YMC America, Inc TRIART C8 120A 5UM 150X2.0MM
SDP
Supplier:  YMC America, Inc TO12S051502WT
Encompass_Preferred
TO12S05-1502WT: Triart C8, 120A, 5um, 150x2.0mm Endcapped Column
Catalog No. 50-394-75