Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triarylsulfonium hexafluoroantimonate salts, mixed

Catalog No. 65402725ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-402-725ML 25 mL
65-402-7100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 65-402-725ML Supplier Sigma Aldrich Supplier No. 65402725ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Triarylsulfonium hexafluoroantimonate salts, mixed are photoinitiators that are used in the cationic polymerization. These photoinitiators are most effective in the range of 210350 nm. They are majorly used to initiate epoxy resins.

Specifications

CAS 109037-75-4
Density 1.410 g/mL (at 25°C)
Boiling Point >220°C
Refractive Index n20/D 1.502
MDL Number MFCD03411230
Quantity 25 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.