Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triarylsulfonium hexafluorophosphate salts, mixed

Catalog No. 407216100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-721-6100ML 100 mL
40-721-625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 40-721-6100ML Supplier Sigma Aldrich Supplier No. 407216100ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Mixed triarylsulfonium hexafluorophosphate salts are cationic photoinitiators which are used as ultraviolet curing agents that have bifunctional cross-linking monomers.It can be mixed with propylene glycol monomethyl ether acetate (PGMA) that may be blended with bisphenol F resin which can be used in the fabrication of UV curable nano-imprinted epoxy resist. It can also be used as a photo-initiator for photo-polymerization of epoxy/acrylate composites which find applications in UV protective coating on electronic devices.

Specifications

CAS 109037-77-6
Density 1.32 g/mL (at 25°C)
Refractive Index n20/D 1.5 (literature)
MDL Number MFCD03412152
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.