Learn More
AdipoGen Triazamate
SDP
Catalog No. 502559992
Encompass_Preferred
Chemical. CAS 112143-82-5. Formula C13H22N4O3S. MW 314.4. Synthetic. Insecticide, which is a plantsystemic aphicide. Inhibits acetylcholinesterase AChE. Specific to aphids. It is safe to beneficial insects and mites. Can be used as a reference compound.
Catalog No. 50-255-9992 Supplier AdipoGen Supplier No. CDXT0123M100
Chemical. CAS 112143-82-5. Formula C13H22N4O3S. MW 314.4. Synthetic. Insecticide, which is a plantsystemic aphicide. Inhibits acetylcholinesterase AChE. Specific to aphids. It is safe to beneficial insects and mites. Can be used as a reference compound.