Learn More
AdipoGen Triazamate
SDP
Supplier:  AdipoGen CDXT0123M500
Encompass_Preferred
Chemical. CAS 112143-82-5. Formula C13H22N4O3S. MW 314.4. Synthetic. Insecticide, which is a plantsystemic aphicide. Inhibits acetylcholinesterase AChE. Specific to aphids. It is safe to beneficial insects and mites. Can be used as a reference compound.
Catalog No. 50-255-9991