Learn More
Cayman Chemical TrIazavIrIn 50mg
SDP
Catalog No. 502913030
Encompass_Preferred
An antiviral agent; inhibits lung viral accumulation in a mouse model of H5N1 influenza A infection at 1 mg/kg; inhibits platelet aggregation in a rat model of LPS-induced hypercytokinemia at 20 mg/kg
Catalog No. 50-291-3030 Supplier Cayman Chemical Supplier No. 3605250 MG
An antiviral agent; inhibits lung viral accumulation in a mouse model of H5N1 influenza A infection at 1 mg/kg; inhibits platelet aggregation in a rat model of LPS-induced hypercytokinemia at 20 mg/kg