Learn More
AdipoGen Triaziflam
SDP
Supplier:  AdipoGen CDXT0124M025
Encompass_Preferred
Chemical. CAS 131475-57-5. Formula C17H24FN5O. MW 333.4. Synthetic. Triazine herbicide. Mode of action is involves the inhibition of the photosystem II electron transport, as well as the inhibition of cellulose biosynthesis. Compound can be used as analytical reference material.
Catalog No. 50-255-9990