Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Triazinetriethanol | 4719-04-4 | MFCD01678788 | ≥96.0% | 219.28 g/mol | C9H21N3O3 | 250 G
SDP

Catalog No. 5000302482
Encompass_Preferred

Triazinetriethanol (CAS 4719-04-4) is a volatile reagent-grade hexahydro-1,3,5-tris(hydroxyethyl)triazine used in analytical and materials research. It is supplied for laboratory research use and has been reported as a volatile component in internal can coatings for food and beverages.

  • High purity (≥96.0%).
  • Molecular formula C9H21N3O3; molecular weight 219.28 g/mol.
  • Reported as a volatile compound in internal can coatings.
  • Provided in multiple pack sizes including 250 g, 500 g, and 1 kg.
  • Intended for research and analytical applications.
  • Available as in-stock reagent for laboratory use.

Catalog No. 50-003-02482 Supplier Medchemexpress LLC Supplier No. 250GHYW018156
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Triazinetriethanol (CAS 4719-04-4) is a volatile reagent-grade hexahydro-1,3,5-tris(hydroxyethyl)triazine used in analytical and materials research. It is supplied for laboratory research use and has been reported as a volatile component in internal can coatings for food and beverages.

  • High purity (≥96.0%).
  • Molecular formula C9H21N3O3; molecular weight 219.28 g/mol.
  • Reported as a volatile compound in internal can coatings.
  • Provided in multiple pack sizes including 250 g, 500 g, and 1 kg.
  • Intended for research and analytical applications.
  • Available as in-stock reagent for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.