Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Triazolo[1,5-c]pyrimidine-2-sulfonamide, N-(2,6-dichlorophenyl)-5-ethoxy-7-fluoro- | 145701-21-9 | 98.20% | 250 MG
SDP

Catalog No. 5000343023
Encompass_Preferred

Diclosulam (XDE 564) is a highly pure herbicide, with a purity of 98.20%. It exhibits a Ki value of <32 nM and MIC and MIC50 values of 6.25, 1.40, 12.5, and 3.01 μM against CBS10913 and CBS12373 strains. This compound is intended for research use only.

  • Acts as a herbicide.
  • Features a molecular formula of C13H10Cl2FN5O3S.
  • Has a molecular weight of 406.22.
  • Appears as a white to off-white solid.
  • Soluble in DMSO (100 mg/mL) and corn oil (≥ 2.5 mg/mL).
  • Powder storage: -20°C for 3 years, 4°C for 2 years.
  • In solvent storage: -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-003-43023 Supplier Medchemexpress LLC Supplier No. 250MGHY12852
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Diclosulam (XDE 564) is a highly pure herbicide, with a purity of 98.20%. It exhibits a Ki value of <32 nM and MIC and MIC50 values of 6.25, 1.40, 12.5, and 3.01 μM against CBS10913 and CBS12373 strains. This compound is intended for research use only.

  • Acts as a herbicide.
  • Features a molecular formula of C13H10Cl2FN5O3S.
  • Has a molecular weight of 406.22.
  • Appears as a white to off-white solid.
  • Soluble in DMSO (100 mg/mL) and corn oil (≥ 2.5 mg/mL).
  • Powder storage: -20°C for 3 years, 4°C for 2 years.
  • In solvent storage: -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.