Learn More
Aviva Systems Biology Corp TRIB1 Antibody - C-terminal region (ARP52413_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52413_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIB1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human TRIB1. Alternate names: C8FW, GIG2, SKIP1, TRB1ApplicationWB, IHC. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-285-714