Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tribasic Calcium Phosphate, Powder, NF, 34-40%, Spectrum™ Chemical
SDP

Catalog No. 18607537 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
100 lb.
Packaging:
Fiber Drum
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-537 100 lb. Fiber Drum
18-607-536 12 kg Fiber Drum
18-607-535 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-607-537 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CA165100LBBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

CA165, 12167-74-7, Ca5(OH)(PO4)3

Spectrum™ Chemical Tribasic Calcium Phosphate, Powder, NF is a widely used pharmaceutical excipient as binder and filler in solid oral dosage forms which include compressed tablets and hard gelatin capsules. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 12167-74-7
Molecular Formula Ca3O8P2
Molecular Weight (g/mol) 310.17
MDL Number MFCD00010904
InChI Key QORWJWZARLRLPR-UHFFFAOYSA-H
IUPAC Name tricalcium diphosphate
SMILES [Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O

Specifications

CAS Max % 1
Quantity 100 lb.
Ignition Residue 0.08
Packaging Fiber Drum
Percent Purity 34 to 40%
Grade NF
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.