Learn More
Cayman Chemical TrIbenosIde 50mg
SDP
Catalog No. 502946489
Encompass_Preferred
A vasoprotective agent; inhibits agonist-induced contractions in various isolated porcine venous preparations from 10-30 UG/ml; inhibits compound 48/80-induced histamine release in isolated rat peritoneal cells (EC50 = 10 UG/ml); reduces decreases in proteoglycan levels and the severity of cartilage destruction in the inflamed joints in a rabbit model of surgically induced osteoarthritis at 100 and 200 mg/kg per day
Catalog No. 50-294-6489 Supplier Cayman Chemical Supplier No. 3349850 MG
A vasoprotective agent; inhibits agonist-induced contractions in various isolated porcine venous preparations from 10-30 UG/ml; inhibits compound 48/80-induced histamine release in isolated rat peritoneal cells (EC50 = 10 UG/ml); reduces decreases in proteoglycan levels and the severity of cartilage destruction in the inflamed joints in a rabbit model of surgically induced osteoarthritis at 100 and 200 mg/kg per day