Learn More
AdipoGen Tribenuron methylester
SDP
Supplier:  AdipoGen CDXT0119M100
Encompass_Preferred
Chemical. CAS 101200-48-0. Formula C15H17N5O6S. MW 395.39. Tribenuron-methyl is the methyl ester of the herbicide tribenuron and also the form in which tribenuron is used in pesticides and belongs to the group of sulfonylurea herbicides. Tribenuron-methyl is a systemic herbicide that is absorbed by the roots and leaves and then spreads throughout the plant. Like other sulphonylurea herbicides, tribenuron-methyl inhibits the cell division of the plants by disrupting the action of the enzyme acetolactate synthase ALS. Can be used as reference compound.
Catalog No. 50-255-9996