Learn More
Selleck Chemical LLC Tribenzagan Hydrochloride
SDP
Supplier:  Selleck Chemical LLC S54831G
Encompass_Preferred
Tribenzagan Hydrochloride (Trimethobenzamide) is an orally available antiemetic agent used in the therapy of nausea and vomiting associated with medications and gastrointestinal viral and other illnesses
Catalog No. 50-000-03579