Learn More
TRIBIOSCIENCE INC Alenphosome - A - Alendronate Liposomes (Anionic), 2 ML
SDP
Catalog No. NC3990371
Encompass
Specifications: Active: alendronate sodium (CAS 121268-17-5), 5.0 mg/mL; Volume: 2.0 mL; Lipid composition: phosphatidylcholine, phosphatidylglycerol and cholesterol
Catalog No. NC3990371 Supplier TRIBIOSCIENCE INC Supplier No. F70103AA
Specifications: Active: alendronate sodium (CAS 121268-17-5), 5.0 mg/mL; Volume: 2.0 mL; Lipid composition: phosphatidylcholine, phosphatidylglycerol and cholesterol