Learn More
TRIBIOSCIENCE INC 2% Bovine Serum Albumin (BSA) Solution in PBS, 50ML
SDP
Catalog No. NC2221814
Encompass
The BSA Solution is formulated with bovine serum albumin Fraction V in 1x PBS (pH 7.0-7.4). It contains 2% bovine serum albumin Fraction V (20 mg/mL) in the buffer. The solution can be used for in vitro assays of molecular biology and protein assay.
This item is not returnable. View return policy
Catalog No. NC2221814 Supplier TRIBIOSCIENCE INC Supplier No. TBS5048
The BSA Solution is formulated with bovine serum albumin Fraction V in 1x PBS (pH 7.0-7.4). It contains 2% bovine serum albumin Fraction V (20 mg/mL) in the buffer. The solution can be used for in vitro assays of molecular biology and protein assay.