Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tribromoacetaldehyde 97%, Thermo Scientific™

Catalog No. AC139290250
Change view
Click to view available options
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC139290250 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC139290250 Supplier Thermo Scientific Chemicals Supplier No. 139290250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 115-17-3
Density 2.665g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C2HBr3O
Refractive Index 1.5845 to 1.5865 (20°C, 589nm)
Linear Formula CBr3CHO
Quantity 25mL
MDL Number MFCD00006961
Beilstein 01, 626
Merck Index 15, 1390
Synonym tribromoacetaldehyde, bromal, acetaldehyde, tribromo, unii-w2wdi7648e, acetaldehyde,2,2,2-tribromo, acetaldehyde, 2,2,2-tribromo, bromal 6ci, tribromo, tribromoethanal, bromal, redist.
InChI Key YTGSYRVSBPFKMQ-UHFFFAOYSA-N
SMILES C(=O)C(Br)(Br)Br
IUPAC Name 2,2,2-tribromoacetaldehyde
Molecular Weight (g/mol) 280.741
PubChem CID 8256
Formula Weight 280.73
Percent Purity ≥96.0%
Chemical Name or Material Tribromoacetaldehyde, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-067-9
RTECSNumber AB3237500
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.