Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tribromoacetic Acid 98.0+%, TCI America™
SDP

Catalog No. T0351500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0351500G 500 g
T035125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0351500G Supplier TCI America Supplier No. T0351500G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 75-96-7
Molecular Formula C2HBr3O2
Molecular Weight (g/mol) 296.74
MDL Number MFCD00004167
InChI Key QIONYIKHPASLHO-UHFFFAOYSA-N
Synonym tribromoacetic acid, acetic acid, tribromo, tribromacetic acid, unii-71ktl99qjx, ccris 7887, 71ktl99qjx, acetic acid, 2,2,2-tribromo, snpljqbkdvidtaetp@, dsstox_cid_1668, acmc-209p0y
PubChem CID 6415
IUPAC Name 2,2,2-tribromoacetic acid
SMILES C(=O)(C(Br)(Br)Br)O

Specifications

Melting Point 131°C
Boiling Point 245°C
Quantity 500 g
UN Number 3261
Formula Weight 296.74
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tribromoacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.