Learn More
Medchemexpress LLC Tribromoethyl alcohol | 75-80-9 | 500 MG
SDP
Catalog No. 5000412774
Encompass_Preferred
Tribromoethyl alcohol, also known as 2,2,2-Tribromoethanol, is primarily used as an anesthetic for laboratory animals, especially rodents, in preparation for surgical procedures. The datasheet provides methods for preparing solutions for in vivo administration, including a 2.5% solution using tert-amyl alcohol and sterile saline, injected intraperitoneally.
  • Used as an anesthetic for laboratory animals.
  • Suitable for rodents.
  • Instructions for in vivo administration.
  • Can be prepared as a 2.5% solution.
Catalog No. 50-004-12774 Supplier Medchemexpress LLC Supplier No. HYB1372500MG
Tribromoethyl alcohol, also known as 2,2,2-Tribromoethanol, is primarily used as an anesthetic for laboratory animals, especially rodents, in preparation for surgical procedures. The datasheet provides methods for preparing solutions for in vivo administration, including a 2.5% solution using tert-amyl alcohol and sterile saline, injected intraperitoneally.
  • Used as an anesthetic for laboratory animals.
  • Suitable for rodents.
  • Instructions for in vivo administration.
  • Can be prepared as a 2.5% solution.