Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α,α,α-Tribromoquinaldine, 98+%, Thermo Scientific™

Catalog No. AC174150500
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC174150500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC174150500 Supplier Thermo Scientific Chemicals Supplier No. 174150500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 613-53-6
Molecular Formula C10H6Br3N
Molecular Weight (g/mol) 379.88
MDL Number MFCD00006755
InChI Key UDYYQHILRSDDMP-UHFFFAOYSA-N
PubChem CID 69178
IUPAC Name 2-(tribromomethyl)quinoline
SMILES BrC(Br)(Br)C1=CC=C2C=CC=CC2=N1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 127°C to 131°C
Color Brown-Orange or Cream
Infrared Spectrum Authentic
Assay Percent Range 98+%
Quantity 50 g
Beilstein 20, II, 243
Synonym 2-tribromomethyl quinoline, quinoline, 2-tribromomethyl, alpha,alpha,alpha-tribromoquinaldine, 2-alpha,alpha,alpha-tribromomethyl quinoline, .alpha.,.alpha.,.alpha.-tribromoquinaldine, acmc-209mrr, 2-tribrommethyl-chinolin, 2-tribromo-methyl-quinoline, quinoline,2-tribromomethyl, udyyqhilrsddmp-uhfffaoysa
Formula Weight 379.88
Percent Purity ≥98.0%
Physical Form Crystalline Powder
Chemical Name or Material α, α, α-Tribromoquinaldine, 98+%
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-347-1
RTECSNumber VC3250750
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.