Learn More
Cayman Chemical TrIbulosIdehydrate 10mg
SDP
Catalog No. 502948050
Encompass_Preferred
A flavonoid; active against M. madagascariense and M. indicus pranii (MICs = 0.8 and 1.6 mg/ml); scavenges DPPH radicals at 63.7-75.2 mg/ml
Catalog No. 50-294-8050 Supplier Cayman Chemical Supplier No. 3321810 MG
A flavonoid; active against M. madagascariense and M. indicus pranii (MICs = 0.8 and 1.6 mg/ml); scavenges DPPH radicals at 63.7-75.2 mg/ml