Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tributyl(1-Ethoxyvinyl)Tin

Catalog No. 2751235G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-512-35G 5 g
27-512-3100G 100 g
27-512-325G 25 g
27-512-31G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Danger
Catalog No. 27-512-35G Supplier Sigma Aldrich Supplier No. 2751235G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

This vinylstannane undergoes Stille coupling with a vinyl triflate, giving, after hydrolysis, an +a,+a-unsaturated ketone, thus acting as an acetyl anion equivalent.Reagent employed in palladium-catalyzed coupling reactions with sulfoxides, +a-haloethers, and chlorocyclobutenones. Also used to convert acid chlorides to +a-oxygenated enones.

Specifications

CAS 97674-02-7
Density 1.069 g/mL at 25°C
Boiling Point 85°C to 86°C (0.1 mmHg, lit.)
Molecular Formula C16H34OSn
Linear Formula [CH3(CH2)3]3SnC(OC2H5)=CH2
Refractive Index n20/D 1.476
MDL Number MFCD00010240
Quantity 5 g
Molecular Weight (g/mol) 361.15 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.