Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tributyl(1-Propynyl)Tin

Catalog No. 4998625G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-986-25G 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 49-986-25G Supplier Sigma Aldrich Supplier No. 4998625G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tributyl(1-propynyl)tin can be used:As a starting material in the synthesis of phenyl-substituted pyrimidine-dione by reacting with ethyl isocyanate via cycloaddition and Stille coupling reactions.In the synthesis of 18F labeled triazolo quinoline derivative, a positron emission tomography (PET) ligand used in the in vivo imaging of metabotropic glutamate receptor type 1.In one of the key synthetic steps for the synthesis of 4-(1-aryltriazol-4-yl)-tetrahydropyridines as potent mGluR1 antagonists.

Specifications

CAS 308796-14-7
Density 0.965 g/mL at 25°C
Boiling Point 65°C
Molecular Formula C8H9BrZn
Refractive Index n20/D 1.483
Linear Formula C6H5CH2CH2ZnBr
MDL Number MFCD01311437
Quantity 50 mL
Molecular Weight (g/mol) 250.45 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.