Learn More
Medchemexpress LLC Tributyl citrate | 77-94-1 | 98.3% | 360.45 | 100 G
SDP
Catalog No. 5000454217
Encompass_Preferred
Tributyl citrate is a citric acid ester used as a pharmaceutical excipient and plasticizer. Pharmaceutical excipients are inactive ingredients that enhance stability, solubility, and processability of drugs, and can influence the absorption, distribution, metabolism, and elimination (ADME) processes of co-administered medications.
  • Citric acid ester
  • Used as a pharmaceutical excipient
  • Functions as a plasticizer
  • Improves drug stability and solubility
  • Enhances drug processability
  • Affects ADME processes of co-administered drugs
Catalog No. 50-004-54217 Supplier Medchemexpress LLC Supplier No. HYW013762100G
Tributyl citrate is a citric acid ester used as a pharmaceutical excipient and plasticizer. Pharmaceutical excipients are inactive ingredients that enhance stability, solubility, and processability of drugs, and can influence the absorption, distribution, metabolism, and elimination (ADME) processes of co-administered medications.
  • Citric acid ester
  • Used as a pharmaceutical excipient
  • Functions as a plasticizer
  • Improves drug stability and solubility
  • Enhances drug processability
  • Affects ADME processes of co-administered drugs