Learn More
Medchemexpress LLC Tributyl citrate | 77-94-1 | 98.3% | 360.45 | 250 G
SDP
Supplier:  Medchemexpress LLC 250GHYW013762
Encompass_Preferred
Tributyl citrate is a citric acid ester that serves as an excipient, specifically as a plasticizer. Pharmaceutical excipients are typically inactive ingredients in pharmaceutical preparations, which can enhance the stability, solubility, and processability of the preparations. Additionally, they can influence the absorption, distribution, metabolism, and elimination (ADME) processes of co-administered drugs.
  • Improves the stability of pharmaceutical preparations.
  • Enhances the solubility of pharmaceutical preparations.
  • Improves the processability of pharmaceutical preparations.
  • Affects the ADME processes of co-administered drugs.
Catalog No. 50-004-51289