Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tributyl O-Acetylcitrate 97.0+%, TCI America™
SDP

Catalog No. A082225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A082225ML 25 mL
A0822500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A082225ML Supplier TCI America Supplier No. A082225ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 77-90-7
Molecular Formula C20H34O8
Molecular Weight (g/mol) 402.48
MDL Number MFCD00043554
InChI Key QZCLKYGREBVARF-UHFFFAOYSA-N
Synonym O-Acetylcitric Acid Tributyl Ester
PubChem CID 6505
IUPAC Name 1,2,3-tributyl 2-(acetyloxy)propane-1,2,3-tricarboxylate
SMILES CCCCOC(=O)CC(CC(=O)OCCCC)(OC(C)=O)C(=O)OCCCC

Specifications

Melting Point -80°C
Color Yellow
Boiling Point 343°C
Quantity 25 mL
Formula Weight 402.48
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Tributyl O-Acetylcitrate
EINECSNumber (2)-1327
RTECSNumber TZ8330000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.