Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tributyl Phosphate, 98.5%+, Reagents

Catalog No. 01336914
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-914 4 L
01-336-917 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-336-914 Supplier Reagents Holdings Llc Supplier No. C23415004A
Add to Cart
Add to Cart

Tributyl Phosphate, 98.5%+, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 126-73-8
Molecular Formula C12H27O4P
Molecular Weight (g/mol) 266.32
InChI Key STCOOQWBFONSKY-UHFFFAOYSA-N
Synonym Tri-n-butyl Phosphate, Butyl Phosphate
IUPAC Name tributyl phosphate
SMILES CCCCOP(=O)(OCCCC)OCCCC

Specifications

Boiling Range 289°C
Melting Point -80°C
Color Colorless to Slight Yellow
Boiling Point 289°C
Melting Range -80°C
Packaging Poly Bottle
Quantity 4 L
Specific Gravity 0.98
Grade Reagent
Physical Form Liquid
Chemical Name or Material Tributyl Phosphate, 98.5%+
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.