Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tributyl phosphate-d27 | 61196-26-7 | MFCD01073491 | 98.2% | 293.48 g/mol | C12D27O4P | 100 MG
SDP

Catalog No. 5000227677
Encompass_Preferred

Deuterium-labeled tributyl phosphate (d27) supplied as a compact reference standard for use as a tracer and internal standard in quantitative nuclear magnetic resonance (NMR), gas chromatography-mass spectrometry (GC-MS), and liquid chromatography-mass spectrometry (LC-MS) analyses. For research use only; follow recommended storage to maintain stability.

  • Deuterium labeled (d27) isotopologue.
  • Intended as tracer and internal standard for NMR, GC-MS, and LC-MS.
  • Molecular weight 293.48 g/mol; formula C12D27O4P.
  • Provided as a 100 mg quantity; aliquot and avoid repeated freeze-thaw.
  • Purity reported around 98.2% (lot-dependent); verify certificate of analysis.

Catalog No. 50-002-27677 Supplier Medchemexpress LLC Supplier No. HY117600S100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Deuterium-labeled tributyl phosphate (d27) supplied as a compact reference standard for use as a tracer and internal standard in quantitative nuclear magnetic resonance (NMR), gas chromatography-mass spectrometry (GC-MS), and liquid chromatography-mass spectrometry (LC-MS) analyses. For research use only; follow recommended storage to maintain stability.

  • Deuterium labeled (d27) isotopologue.
  • Intended as tracer and internal standard for NMR, GC-MS, and LC-MS.
  • Molecular weight 293.48 g/mol; formula C12D27O4P.
  • Provided as a 100 mg quantity; aliquot and avoid repeated freeze-thaw.
  • Purity reported around 98.2% (lot-dependent); verify certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.