Learn More
Medchemexpress LLC Tributyl phosphate-d27 | 61196-26-7 | MFCD01861663 | 100.0% | 293.48 g·mol⁻¹ | C12D27O4P | 25 MG
SDP
Catalog No. 5000226809
Encompass_Preferred
Tributyl phosphate-d27 is the deuterium-labeled analog of tributyl phosphate supplied as a high-purity isotopically labeled reagent for use as a tracer and as an internal standard in quantitative analytical methods. It is intended for laboratory research use and available in small, analytical-scale packages suitable for method development and validation.
  • Deuterium-labeled internal standard for NMR, GC-MS, and LC-MS.
  • High chemical purity (GC) supports accurate quantitation.
  • Isotopically enriched to enable tracer and mass-shift applications.
  • Small packaging options ideal for analytical method development.
  • Stable under recommended storage conditions for labeled reagents.
Catalog No. 50-002-26809 Supplier Medchemexpress LLC Supplier No. HY117600S25MG
Tributyl phosphate-d27 is the deuterium-labeled analog of tributyl phosphate supplied as a high-purity isotopically labeled reagent for use as a tracer and as an internal standard in quantitative analytical methods. It is intended for laboratory research use and available in small, analytical-scale packages suitable for method development and validation.
  • Deuterium-labeled internal standard for NMR, GC-MS, and LC-MS.
  • High chemical purity (GC) supports accurate quantitation.
  • Isotopically enriched to enable tracer and mass-shift applications.
  • Small packaging options ideal for analytical method development.
  • Stable under recommended storage conditions for labeled reagents.