Learn More
Medchemexpress LLC Tributyl phosphate-d27 | 61196-26-7 | MFCD01861663 | 98.2% | 293.48 g/mol | C12D27O4P | 5 MG
SDP
Catalog No. 5000307965
Encompass_Preferred
Tributyl phosphate-d27 is the deuterium-labeled analogue of tributyl phosphate used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It is provided for research use and supplied in small, precisely measured quantities.
  • Deuterium-labeled internal standard for NMR, GC-MS, and LC-MS.
  • Suitable as a tracer in analytical and pharmacokinetic studies.
  • High isotopic enrichment with documented purity and COA availability.
  • Available in small reagent quantities suitable for analytical workflows.
  • Stable when stored under recommended conditions for long-term use.
Catalog No. 50-003-07965 Supplier Medchemexpress LLC Supplier No. HY117600S5MG
Tributyl phosphate-d27 is the deuterium-labeled analogue of tributyl phosphate used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It is provided for research use and supplied in small, precisely measured quantities.
  • Deuterium-labeled internal standard for NMR, GC-MS, and LC-MS.
  • Suitable as a tracer in analytical and pharmacokinetic studies.
  • High isotopic enrichment with documented purity and COA availability.
  • Available in small reagent quantities suitable for analytical workflows.
  • Stable when stored under recommended conditions for long-term use.