Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tributyl phosphate, For extraction analysis (GC), ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012867 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2867 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2867 Supplier MilliporeSigma Supelco Supplier No. 00675500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

MDL Number MFCD00009436
Synonym TBP; TBPA

Specifications

Melting Point -79°C
CAS 126-73-8
Density 0.979 g/mL (at 25°C)
Boiling Point 180°C to 183°C (22 mmHg)
Flash Point 145°C
Ignition Point 770°F
Linear Formula (CH3(CH2)3 O)3 PO
Quantity 500 mL
Vapor Density 9.2 (vs. air)
Vapor Pressure 7.3 mmHg (150°C)
Formula Weight 266.31
Percent Purity ≥99.0% (GC)
Grade For extraction analysis
Show More Show Less
RTECSNumber TC7700000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.