Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tributyl(Vinyl)Tin

Catalog No. 503081494 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
250 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-308-1494 1 g
27-143-825G 25 g
27-143-85G 5 g
27-143-8250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Danger
Catalog No. 50-308-1494 Supplier Sigma Aldrich Supplier No. 2714381G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Vinyl nucleophile for bromoacetylenes and bromoaromatics.

Specifications

CAS 7486-35-3
Density 1.085 g/mL at 25°C
Boiling Point 104°C to 106°C (3.5 mmHg, lit.)
Molecular Formula C14H30Sn
Refractive Index n20/D 1.478
Linear Formula CH2=CHSn[CH3(CH2)3]3
MDL Number MFCD00009421
Quantity 1 g
Synonym Tributyl(vinyl)stannane; Tributylstannylethylene; Vinyltributylstannane
Molecular Weight (g/mol) 317.1 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.