Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tributylamine

Catalog No. 47131325ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
250 mL
2.5 L
18 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-131-325ML 25 mL
47-131-318L 18 L
47-131-325L 2.5 L
47-131-3250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 47-131-325ML Supplier Sigma Aldrich Supplier No. 47131325ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tributylamine

Specifications

CAS 102-82-9
Melting Point -70°C
Density 0.778 g/mL at 25°C
Boiling Point 216°C
Molecular Formula C12H27N
Refractive Index n20/D 1.428
Linear Formula [CH3(CH2)3]3N
MDL Number MFCD00009431
Quantity 25 mL
Molecular Weight (g/mol) 185.35 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.