Learn More
Medchemexpress LLC Tributylbenzylammonium bromide | 25316-59-0 | 98.0% | 356.38 | 1 KG
SDP
Catalog No. 5000452814
Encompass_Preferred
Tributylbenzylammonium bromide is a quaternary ammonium salt consisting of a positively charged N-benzyl-N,N-dibutylbutan-1-amine cation and a negatively charged bromide anion. It is commonly used as a surfactant and emulsifier in products such as detergents, fabric softeners, and personal care items. It also possesses antimicrobial properties, making it suitable for use as a disinfectant or antiseptic in certain products. For research use only, not for sale to patients. This biochemical reagent can be used as a biological material or organic compound for life science-related research.
  • Can be used as a biological material
  • Can be used as an organic compound for life science-related research
Catalog No. 50-004-52814 Supplier Medchemexpress LLC Supplier No. HYW0111221KG
Tributylbenzylammonium bromide is a quaternary ammonium salt consisting of a positively charged N-benzyl-N,N-dibutylbutan-1-amine cation and a negatively charged bromide anion. It is commonly used as a surfactant and emulsifier in products such as detergents, fabric softeners, and personal care items. It also possesses antimicrobial properties, making it suitable for use as a disinfectant or antiseptic in certain products. For research use only, not for sale to patients. This biochemical reagent can be used as a biological material or organic compound for life science-related research.
  • Can be used as a biological material
  • Can be used as an organic compound for life science-related research