Learn More
Medchemexpress LLC Tributylbenzylammonium bromide | 25316-59-0 | 98.0% | 356.38 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW011122100G
Encompass_Preferred
Tributylbenzylammonium bromide is a quaternary ammonium salt used as a surfactant and emulsifier in various applications, including detergents, fabric softeners, and personal care products. It also possesses antimicrobial properties and is utilized as a disinfectant or antiseptic in certain products.
  • Biochemical reagent for life science-related research.
  • Suitable for laboratory use.
  • Available in various pack sizes.
Catalog No. 50-004-51804