Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tributylmethylammonium Chloride Solution

Catalog No. 255165500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-516-5500G 500 g
25-516-5100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 25-516-5500G Supplier Sigma Aldrich Supplier No. 255165500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tributylmethylammonium Chloride Solution

Specifications

CAS 56375-79-2
Density 0.964 g/mL at 25°C
Boiling Point 152°C
Molecular Formula C13H30ClN
Refractive Index n20/D 1.4682
Linear Formula CH3N[CH3(CH2)3]3Cl
MDL Number MFCD00011847
Quantity 500 g
Synonym Methyltributylammonium chloride solution
Molecular Weight (g/mol) 235.84 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.